Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252457.1 | NP_001239386.1 | 6 | 790 | |
NM_019693.3 | NP_062667.1 | 6 | 878 | ||
GenBank mRNA | AF118128.1 | - | 5 | 644 | |
AK051034.1 | - | 6 | 811 | ||
AK088867.1 | - | 6 | 796 | ||
AK171507.1 | - | 6 | 820 | ||
AK194985.1 | - | 2 | 195 | ||
AK195134.1 | - | 2 | 195 | ||
AK217230.1 | - | 2 | 165 | ||
AY255786.1 | - | 5 | 770 | ||
AY255787.1 | - | 5 | 770 | ||
BC011067.1 | - | 6 | 786 | 3498520 | |
BC024859.1 | - | 5 | 2014 | 5359825 |