Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019780.1 | NP_062754.1 | 3 | 409 | |
GenBank mRNA | AB578899.1 | - | 2 | 181 | |
AB578900.1 | - | 2 | 181 | ||
AB578901.1 | - | 2 | 181 | ||
AF193794.1 | - | 3 | 409 | ||
AK004103.1 | - | 4 | 426 | ||
AK008248.1 | - | 4 | 429 | ||
AK036426.1 | - | 3 | 397 | ||
AK150330.1 | - | 3 | 419 | ||
BC005663.1 | - | 3 | 398 | 2645432 |