Gene Transcripts

Gene Symbol
Ube2l6
Entrez Gene ID
56791
Gene Name
ubiquitin-conjugating enzyme E2L 6
Gene Aliases
2810489I21Rik, RIG-B, UBCH8, Ubce8, Ubcm8
Chromosome Location
Chr. 2: 84798828 - 84810003 on Build GRCm38
UniGene ID
Mm.38261
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019949.2 NP_064333.2 4 940
GenBank mRNA AF159230.1 - 4 919
AK010942.1 - 4 919
AK013452.1 - 4 930
AK152010.1 - 4 920
AK152066.1 - 4 892
AK152113.1 - 4 941
AK168695.1 - 4 878
AK171629.1 - 4 889
AK206200.1 - 1 21
BC008238.1 - 4 908 2651702

View Full Product Details