Gene Transcripts

Gene Symbol
Nfu1
Entrez Gene ID
56748
Gene Name
NFU1 iron-sulfur cluster scaffold homolog (S. cerevisiae)
Gene Aliases
0610006G17Rik, CGI-33, Hirip5
Chromosome Location
Chr. 6: 87009291 - 87028461 on Build GRCm38
UniGene ID
Mm.23809
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170591.1 NP_001164062.1 7, 8 796
NM_020045.3 NP_064429.2 7, 8 793
GenBank mRNA AJ132616.1 - 7, 8 724
AK002244.1 - 7, 8 742
AK003730.1 - 7, 8 741
AK145558.1 - 7, 8 797
AY335195.1 - 7, 8 700
BC018355.1 - 7, 8 714 3584656

View Full Product Details