Gene Transcripts

Gene Symbol
Wdr46
Entrez Gene ID
57315
Gene Name
WD repeat domain 46
Gene Aliases
2310007I04Rik, Bing4, C78447, C78559
Chromosome Location
Chr. 17: 33940634 - 33949695 on Build GRCm38
UniGene ID
Mm.2437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020603.2 NP_065628.1 4 422
GenBank mRNA AK145068.1 - 4 477
AK216977.1 - 2 83
BC013527.1 - 4 465
BC026431.1 - 1 46
BC046977.1 - 4 422 6315172
KJ650211.1 - 4 407
KJ663716.1 - 4 407

View Full Product Details