Gene Transcripts

Gene Symbol
Ngly1
Entrez Gene ID
59007
Gene Name
N-glycanase 1
Gene Aliases
1110002C09Rik, PNGase, Png1
Chromosome Location
Chr. 14: 16249314 - 16311926 on Build GRCm38
UniGene ID
Mm.258381
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021504.3 NP_067479.2 4, 5 710
GenBank mRNA AF250927.1 - 4, 5 697
AK028248.1 - 4, 5 711
AK148931.1 - 4, 5 713
AK169205.1 - 4, 5 698
BC028961.1 - 4, 5 664 5366023

View Full Product Details