Gene Transcripts

Gene Symbol
Nek7
Entrez Gene ID
59125
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 7
Gene Aliases
2810460C19Rik, AU020186
Chromosome Location
Chr. 1: 138484106 - 138619779 on Build GRCm38
UniGene ID
Mm.143817
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021605.3 NP_067618.1 5 488
GenBank mRNA AF217650.1 - 4 339
AK035502.1 - 5 487
AK078639.1 - 5 497
AK088173.1 - 5 537
AK149308.1 - 5 536
AK165583.1 - 5 466
BC026541.1 - 5 480
BC037697.1 - 5 465 3994871

View Full Product Details