Gene Transcripts

Gene Symbol
Svep1
Entrez Gene ID
64817
Gene Name
sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
Gene Aliases
1110021D17Rik, 4833413O10Rik, AA387071, D430029O09Rik, Polydom
Chromosome Location
Chr. 4: 58042796 - 58206596 on Build GRCm38
UniGene ID
Mm.261024
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022814.2 NP_073725.2 3 1158
GenBank mRNA AF206329.1 - 3 1158
AK035333.1 - 3 1176
AK052699.1 - 3 1130

View Full Product Details