Gene Transcripts

Gene Symbol
Rps19
Entrez Gene ID
20085
Gene Name
ribosomal protein S19
Gene Aliases
Dsk3
Chromosome Location
Chr. 7: 24884376 - 24889806 on Build GRCm38
UniGene ID
Mm.300281
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023133.1 NP_075622.1 6 609
GenBank mRNA AK012052.1 - 5 1266
AK013524.1 - 6 489
AK018725.1 - 5 468
AK028244.1 - 6 485
BC034506.1 - 6 480 3968247
BC086775.1 - 5 471
BC086938.1 - 6 486 30121701

View Full Product Details