Gene Transcripts

Gene Symbol
Prl8a8
Entrez Gene ID
74188
Gene Name
prolactin family 8, subfamily a, member 81
Gene Aliases
1600032B14Rik, AV036230, PLP-Cgamma, Prlpc3
Chromosome Location
Chr. 13: 27507032 - 27513228 on Build GRCm38
UniGene ID
Mm.46090
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023741.2 NP_076230.1 6 625
GenBank mRNA AF230921.1 - 6 611
AF230922.1 - 6 614
AF230923.1 - 7 827
AF466150.1 - 6 626
AK005568.1 - 6 631
AK167369.1 - 6 632
BC100326.1 - 6 619 30787396

View Full Product Details