Gene Transcripts

Gene Symbol
Morf4l1
Entrez Gene ID
21761
Gene Name
mortality factor 4 like 1
Gene Aliases
MORFRG15, MRG15, TEG-189, Tex189, mKIAA4002
Chromosome Location
Chr. 9: 90091669 - 90114820 on Build GRCm38
UniGene ID
Mm.426209
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039147.2 NP_001034236.1 3 349
NM_024431.3 NP_077751.1 3 349
GenBank mRNA AB042241.1 - 3 198
AF319620.1 - 3 197
AF319621.1 - 3 197
AK010528.1 - 3 118
AK011249.1 - 3 226
AK011441.1 - 3 121
AK016414.1 - 3 316
AK146210.1 - 3 184
AK146785.1 - 2 79
AK150925.1 - 3 150
AK152119.1 - 3 126
AK167079.1 - 3 108
AK168327.1 - 3 184
AK168585.1 - 3 129
AK168608.1 - 3 131
AK168674.1 - 3 180
AK169065.1 - 3 181
AK182816.1 - 3 121
AK187780.1 - 3 121
AK190529.1 - 3 121
AK198286.1 - 3 121
AK207086.1 - 3 121
AK207233.1 - 3 121
AK211776.1 - 3 120
AK212639.1 - 3 121
AK212964.1 - 3 121
AK213758.1 - 3 121
AK214297.1 - 3 121
AK215083.1 - 3 121
AK215514.1 - 3 117
AK218105.1 - 3 121
AK218718.1 - 3 121
AK218976.1 - 3 121
AK220441.1 - 3 169
BC003894.1 - 3 201 3593982
BC017619.1 - 3 201 4923923
BC083094.1 - 3 134
BC083118.1 - 3 210 5371036
BC085103.1 - 3 199 6490291
BC092293.1 - 3 213 5068516
BC103783.1 - 3 213 6529024

View Full Product Details