Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025383.3 | NP_079659.1 | 2 | 203 | |
GenBank mRNA | AK003463.1 | - | 2 | 170 | |
AK145874.1 | - | 2 | 191 | ||
AK155871.1 | - | 2 | 191 | ||
AK156566.1 | - | 2 | 180 | ||
AK170987.1 | - | 2 | 188 | ||
AK172538.1 | - | 2 | 173 | ||
AK213029.1 | - | 2 | 95 | ||
AK218572.1 | - | 2 | 112 | ||
BC037069.1 | - | 2 | 202 | 5002765 |