Gene Transcripts

Gene Symbol
Zfand1
Entrez Gene ID
66361
Gene Name
zinc finger, AN1-type domain 1
Gene Aliases
2310008M20Rik, AW048890
Chromosome Location
Chr. 3: 10339953 - 10351338 on Build GRCm38
UniGene ID
Mm.371617
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025512.2 NP_079788.2 8 749
GenBank mRNA AK009240.1 - 8 763
AK017696.1 - 8 759
AK030472.1 - 8 757
AK077873.1 - 8 759
AK081710.1 - 2 3843
BC023352.1 - 8 741
BC025216.1 - 8 741
BC046585.1 - 8 749 6314252
BC089494.1 - 8 761 30504018

View Full Product Details