Gene Transcripts

Gene Symbol
Dopey2
Entrez Gene ID
70028
Gene Name
dopey family member 2
Gene Aliases
0610038M01Rik, 2610510B01Rik, AI503246, C21orf5, mKIAA0933
Chromosome Location
Chr. 16: 93711907 - 93810588 on Build GRCm38
UniGene ID
Mm.23230
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026700.2 NP_080976.1 4 667
NM_027293.1 NP_081569.1 4 667
GenBank mRNA AK002808.1 - 4 597
AK147255.1 - 4 645
AK147536.1 - 3 687
AK156170.1 - 3 1274
AK164839.1 - 4 644
BC075639.1 - 2 179

View Full Product Details