Gene Transcripts

Gene Symbol
Wdfy1
Entrez Gene ID
69368
Gene Name
WD repeat and FYVE domain containing 1
Gene Aliases
1700013B03Rik, 1700120F24Rik, FENS-1, Jr1, WDF1, ZFYVE17, mKIAA1435
Chromosome Location
Chr. 1: 79702262 - 79776047 on Build GRCm38
UniGene ID
Mm.293273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111279.1 NP_001104749.1 1, 2 209
NM_027057.3 NP_081333.1 1, 2 209
GenBank mRNA AK005937.1 - 1, 2 173
AK007215.1 - 1, 2 210
AK157204.1 - 1, 2 168
AK159710.1 - 1, 2 180
AK165554.1 - 1, 2 182
AK220391.1 - 1, 2 143
BC025226.1 - 1, 2 123 5011404

View Full Product Details