Gene Transcripts

Gene Symbol
Med6
Entrez Gene ID
69792
Gene Name
mediator complex subunit 6
Gene Aliases
1500012F11Rik, AU019660, AV213303
Chromosome Location
Chr. 12: 81573564 - 81595071 on Build GRCm38
UniGene ID
Mm.279993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027213.2 NP_081489.2 6 496
GenBank mRNA AK005232.1 - 6 541
AK171784.1 - 6 512
BC013096.1 - 6 496 3710612

View Full Product Details