Gene Transcripts

Gene Symbol
Def6
Entrez Gene ID
23853
Gene Name
differentially expressed in FDCP 6
Gene Aliases
2410003F05Rik, 6430538D02Rik, AV094905, Ibp, Slat, Slat2, Slat6
Chromosome Location
Chr. 17: 28207778 - 28228608 on Build GRCm38
UniGene ID
Mm.204731
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027185.3 NP_081461.2 11 2115
GenBank mRNA AK010356.1 - 3 589
AK038050.1 - 11 2086
AK088460.1 - 11 2064

View Full Product Details