Gene Transcripts

Gene Symbol
Lce1a2
Entrez Gene ID
73722
Gene Name
late cornified envelope 1A2
Gene Aliases
1110004E04Rik, Sprrl2
Chromosome Location
Chr. 3: 92668613 - 92670315 on Build GRCm38
UniGene ID
Mm.279773
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028625.2 NP_082901.1 2 688
GenBank mRNA AK003412.1 - 2 689
BC062142.1 - 2 665
BC080817.1 - 2 665
BC100540.1 - 2 675

View Full Product Details