Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000348.3 | NP_000339.2 | 1, 2 | 360 | |
XM_011533072.2 | XP_011531374.1 | 4, 5 | 505 | ||
GenBank mRNA | AK316597.1 | - | 1, 2 | 330 | |
BC112252.1 | - | 1, 2 | 347 | 8327720 | |
BC113641.1 | - | 1, 2 | 335 | 8322693 | |
EF560740.1 | - | 1, 2 | 328 | ||
M74047.1 | - | 1, 2 | 316 |