Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029365.2 | NP_083641.1 | 7, 8 | 829 | |
GenBank mRNA | AK089940.1 | - | 7, 8 | 833 | |
AK090026.1 | - | 7, 8 | 855 | ||
AK149867.1 | - | 8, 9 | 1101 | ||
AK158238.1 | - | 7, 8 | 890 | ||
AK165760.1 | - | 7, 8 | 875 | ||
BC021333.1 | - | 7, 8 | 864 | 5027401 | |
BC031138.1 | - | 7, 8 | 872 | 4972378 | |
BC071206.1 | - | 6, 7 | 906 | 4925452 |