Gene Transcripts

Gene Symbol
Htra3
Entrez Gene ID
78558
Gene Name
HtrA serine peptidase 3
Gene Aliases
2210021K23Rik, 9530081K03Rik, Prsp, Tasp
Chromosome Location
Chr. 5: 35652026 - 35679813 on Build GRCm38
UniGene ID
Mm.274255
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030127.3 NP_084403.2 8 1630
GenBank mRNA AK035194.1 - 8 1633
AY037300.1 - 8 1415
AY156509.1 - 8 1322
BC138587.1 - 8 1354 8861607
BC138588.1 - 8 1354 8861608

View Full Product Details