Gene Transcripts

Gene Symbol
Sacm1l
Entrez Gene ID
83493
Gene Name
SAC1 (suppressor of actin mutations 1, homolog)-like (S. cerevisiae)
Gene Aliases
SAC1, Sac1p, mKIAA0851
Chromosome Location
Chr. 9: 123529882 - 123592598 on Build GRCm38
UniGene ID
Mm.273671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030692.4 NP_109617.1 20 2894
GenBank mRNA AJ297526.1 - 20 2895
AK122390.1 - 20 2959
AK154759.1 - 20 2994
BC005489.1 - 11 2103

View Full Product Details