Gene Transcripts

Gene Symbol
Tars
Entrez Gene ID
110960
Gene Name
threonyl-tRNA synthetase
Gene Aliases
D15Wsu59e, ThrRS
Chromosome Location
Chr. 15: 11383663 - 11399658 on Build GRCm38
UniGene ID
Mm.286061
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033074.3 NP_149065.2 2 196
GenBank mRNA AK010256.1 - 2 163
AK011146.2 - 2 197
AK031064.1 - 2 160
AK152945.1 - 2 157
AK153314.1 - 2 168
AK166693.1 - 2 163
AK167597.1 - 2 205
AK168969.1 - 2 156
BC055371.1 - 2 182 30060749
BC060176.1 - 139

View Full Product Details