Gene Transcripts

Gene Symbol
NOTCH3
Entrez Gene ID
4854
Gene Name
notch 3
Gene Aliases
CADASIL, CADASIL1, CASIL, IMF2, LMNS
Chromosome Location
Chr. 19: 15159633 - 15200981 on Build GRCh38
UniGene ID
Hs.8546
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000435.2 NP_000426.2 7 1170
XM_005259924.4 XP_005259981.1 7 1109
GenBank mRNA AB209447.1 - 7 1103
U97669.1 - 7 1172

View Full Product Details