Gene Transcripts

Gene Symbol
Tmc2
Entrez Gene ID
192140
Gene Name
transmembrane channel-like gene family 2
Gene Aliases
CWEA2
Chromosome Location
Chr. 2: 130195194 - 130264445 on Build GRCm38
UniGene ID
Mm.219546
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138655.1 NP_619596.1 13 1880
GenBank mRNA AF417581.1 - 13 1880
AY263156.1 - 2 47

View Full Product Details