Gene Transcripts

Gene Symbol
Adig
Entrez Gene ID
246747
Gene Name
adipogenin
Gene Aliases
SMAF-1, Smaf1
Chromosome Location
Chr. 2: 158502612 - 158508198 on Build GRCm38
UniGene ID
Mm.28800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145635.2 NP_663610.1 2 343
GenBank mRNA AK131610.1 - 2 293
AY092026.1 - 2 349
BC054059.1 - 2 291 3373967
DQ223719.1 - 2 212

View Full Product Details