Gene Transcripts

Gene Symbol
Lace1
Entrez Gene ID
215951
Gene Name
lactation elevated 1
Gene Aliases
CG8520
Chromosome Location
Chr. 10: 42312581 - 42478584 on Build GRCm38
UniGene ID
Mm.89646
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145743.2 NP_665686.2 13 2476
GenBank mRNA AF397909.1 - 12 2179
AF520417.1 - 13 2325
AK044595.1 - 13 2352
AK170996.1 - 14 2519
AK207892.1 - 1 175
BC023308.1 - 12 2153
BC025880.1 - 2 1031
BC089595.1 - 13 2326 30118411

View Full Product Details