Gene Transcripts

Gene Symbol
Tspan33
Entrez Gene ID
232670
Gene Name
tetraspanin 33
Gene Aliases
1300010A20Rik, Pen
Chromosome Location
Chr. 6: 29694214 - 29718558 on Build GRCm38
UniGene ID
Mm.225289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146173.3 NP_666285.1 2 380
GenBank mRNA AF276890.1 - 2 379
AK004954.1 - 2 354
AK162631.1 - 1 1975
BC024685.1 - 2 338 3981492

View Full Product Details