Gene Transcripts

Gene Symbol
Slc5a11
Entrez Gene ID
233836
Gene Name
solute carrier family 5 (sodium/glucose cotransporter), member 11
Gene Aliases
2010013B02Rik, Kst1, RKST2, SMIT2, Slc5a10
Chromosome Location
Chr. 7: 123214758 - 123274390 on Build GRCm38
UniGene ID
Mm.211838
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146198.2 NP_666310.1 17 2214
GenBank mRNA AF518726.1 - 15 1635
AK008219.1 - 17 2269
BC031742.1 - 17 2215 4221537

View Full Product Details