Gene Transcripts

Gene Symbol
RPS29
Entrez Gene ID
6235
Gene Name
ribosomal protein S29
Gene Aliases
DBA13, S29
Chromosome Location
Chr. 14: 49576672 - 49586416 on Build GRCh38
UniGene ID
Hs.156367
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030001.2 NP_001025172.1 2 222
NM_001032.4 NP_001023.1 2 222
GenBank mRNA AK026649.1 - 1 182
AK307833.1 - 2 183
BC015974.1 - 2 186 4105424
BC032813.1 - 1 157 5217083
BC035313.1 - 157 3868721
BF128896.1 - 2 129
BQ433530.1 - 2 166
BU942244.1 - 2 225
L31610.1 - 2 163
U14973.1 - 2 182

View Full Product Details