Gene Transcripts

Gene Symbol
Peo1
Entrez Gene ID
226153
Gene Name
progressive external ophthalmoplegia 1 (human)
Gene Aliases
D19Ertd626e, PEO, Twinl
Chromosome Location
Chr. 19: 45006079 - 45012762 on Build GRCm38
UniGene ID
Mm.105585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153796.3 NP_722491.2 1 908
GenBank mRNA AK146244.1 - 1 802
AY059385.1 - 1 370
BC071195.1 - 1 908 5100486

View Full Product Details