Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172300.3 | NP_758504.3 | 4, 5 | 783 | |
GenBank mRNA | AK005524.1 | - | 2, 3 | 334 | |
AK077793.1 | - | 4, 5 | 782 | ||
AK149770.1 | - | 4, 5 | 783 | ||
AK152529.1 | - | 4, 5 | 497 | ||
AK153328.1 | - | 4, 5 | 514 | ||
AK168942.1 | - | 4, 5 | 476 | ||
BC027266.1 | - | 4, 5 | 408 | ||
BC054412.1 | - | 4, 5 | 486 | 2646768 |