Gene Transcripts

Gene Symbol
Dph3
Entrez Gene ID
105638
Gene Name
diphthamine biosynthesis 3
Gene Aliases
DELGIP1, DelgipP1, Desr1, KTI11, Zcsl2
Chromosome Location
Chr. 14: 32080517 - 32085692 on Build GRCm38
UniGene ID
Mm.2519
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047433.2 NP_001040898.1 2 1645
NM_172254.4 NP_758458.1 3 1720
GenBank mRNA AK048097.1 - 3 1610

View Full Product Details