Gene Transcripts

Gene Symbol
Mfsd4
Entrez Gene ID
213006
Gene Name
major facilitator superfamily domain containing 4
Gene Aliases
A230072B04, A930031D07Rik, AI850289
Chromosome Location
Chr. 1: 132022806 - 132068062 on Build GRCm38
UniGene ID
Mm.208445
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114662.1 NP_001108134.1 2 496
NM_172510.4 NP_766098.2 2 496
GenBank mRNA AK038882.1 - 2 389
AK044667.1 - 2 369
AK085484.1 - 2 368
AK162495.1 - 2 400
BC058790.1 - 2 345 6395070

View Full Product Details