Gene Transcripts

Gene Symbol
Ankle1
Entrez Gene ID
234396
Gene Name
ankyrin repeat and LEM domain containing 1
Gene Aliases
8430438L13Rik, Ankrd41, C430003I23, Lem3
Chromosome Location
Chr. 8: 71406005 - 71410543 on Build GRCm38
UniGene ID
Mm.216549
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172756.2 NP_766344.2 6 1131
GenBank mRNA AK049395.1 - 4 1438
AK078830.1 - 5 1361
AK080768.1 - 4 1444
AK083552.1 - 4 1430
AK087444.1 - 4 1449
BC030436.1 - 6 1070
BC094316.1 - 6 1088
BC145659.1 - 2 354
EU184014.1 - 6 1107

View Full Product Details