Gene Transcripts

Gene Symbol
Maob
Entrez Gene ID
109731
Gene Name
monoamine oxidase B
Gene Aliases
6330414K01Rik, MAO-B
Chromosome Location
Chr. X: 16709281 - 16817366 on Build GRCm38
UniGene ID
Mm.241656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172778.2 NP_766366.2 7, 8 867
GenBank mRNA AK031833.1 - 7, 8 851
AK054050.1 - 7, 8 866
BC113182.1 - 7, 8 856 40061991
BC113788.1 - 7, 8 856 40061990

View Full Product Details