Gene Transcripts

Gene Symbol
Peli3
Entrez Gene ID
240518
Gene Name
pellino 3
Gene Aliases
6030441F14Rik, A930011L17, BC028931
Chromosome Location
Chr. 19: 4929717 - 4943127 on Build GRCm38
UniGene ID
Mm.277580
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172835.3 NP_766423.3 2 281
GenBank mRNA AK044418.1 - 2 228
AK135575.1 - 2 230
BC028931.1 - 2 239 5037308
BY128359.1 - 2 281

View Full Product Details