Gene Transcripts

Gene Symbol
Peli3
Entrez Gene ID
240518
Gene Name
pellino 3
Gene Aliases
6030441F14Rik, A930011L17, BC028931
Chromosome Location
Chr. 19: 4929717 - 4943127 on Build GRCm38
UniGene ID
Mm.277580
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172835.3 NP_766423.3 6 852
GenBank mRNA AK044418.1 - 6 799
BC028931.1 - 6 810 5037308

View Full Product Details