Gene Transcripts

Gene Symbol
Manea
Entrez Gene ID
242362
Gene Name
mannosidase, endo-alpha
Gene Aliases
4932703L02Rik
Chromosome Location
Chr. 4: 26324506 - 26346652 on Build GRCm38
UniGene ID
Mm.245602
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172865.2 NP_766453.2 5 1324
GenBank mRNA AK030141.1 - 5 1299
BC067076.1 - 5 1324 30628555

View Full Product Details