Gene Transcripts

Gene Symbol
Tango6
Entrez Gene ID
272538
Gene Name
transport and golgi organization 6
Gene Aliases
AW413431, E330010G16, Tmco7
Chromosome Location
Chr. 8: 106683068 - 106851439 on Build GRCm38
UniGene ID
Mm.87151
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173037.1 NP_766625.1 1, 2 196
GenBank mRNA AK054283.1 - 1, 2 150
AK076373.1 - 1, 2 118
AK077757.1 - 1, 2 182
AK085030.1 - 1, 2 196
AK173250.1 - 1, 2 103
BC066134.1 - 1, 2 126
BC066869.1 - 1, 2 126 30106406
BC069838.1 - 1, 2 126 30106422

View Full Product Details