Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_176933.4 | NP_795907.1 | 4 | 1592 | |
GenBank mRNA | AK053746.1 | - | 4 | 1589 | |
AK080964.1 | - | 4 | 1592 | ||
AK165991.1 | - | 4 | 1909 | ||
BC118941.1 | - | 4 | 1563 | 40092218 | |
BC118942.1 | - | 4 | 1561 | 40092219 |