Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_177082.4 | NP_796056.2 | 3 | 1069 | |
GenBank mRNA | AJ844914.1 | - | 1 | 756 | |
AK030745.1 | - | 2 | 888 | ||
AK053093.1 | - | 3 | 1069 | ||
BC082582.1 | - | 2 | 943 | 30653287 |