Gene Transcripts

Gene Symbol
Vash1
Entrez Gene ID
238328
Gene Name
vasohibin 1
Gene Aliases
AI834978, D930046M13Rik, G630009D10Rik
Chromosome Location
Chr. 12: 86678700 - 86695688 on Build GRCm38
UniGene ID
Mm.216101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177354.4 NP_796328.2 7 2470
GenBank mRNA AB284948.1 - 7 2475
AK090166.1 - 7 2476
BC086688.1 - 7 970 30686635

View Full Product Details