Gene Transcripts

Gene Symbol
Lppr4
Entrez Gene ID
229791
Gene Name
lipid phosphate phosphatase-related protein type 4
Gene Aliases
A330086D10, D3Bwg0562e, PRG-1, mKIAA0455
Chromosome Location
Chr. 3: 117319145 - 117360876 on Build GRCm38
UniGene ID
Mm.140138
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177664.5 NP_808332.3 1 795
GenBank mRNA AF541279.1 - 1 796
AK046782.1 - 1 796
AK079635.1 - 1 129
AK129149.1 - 1 391
AY266266.1 - 1 796
AY266267.1 - 1 796
BC082323.1 - 1 325
BC137701.1 - 1 390 8860720
BC137702.1 - 1 391 8860721

View Full Product Details