Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290519.1 | NP_001277448.1 | 5, 6 | 421 | |
NM_001290520.1 | NP_001277449.1 | 3, 4 | 344 | ||
NM_177576.3 | NP_808244.1 | 4, 5 | 448 | ||
GenBank mRNA | AK052771.1 | - | 4, 5 | 449 | |
AK132922.1 | - | 5, 6 | 422 | ||
BC109334.2 | - | 3, 4 | 258 | 40051782 | |
HQ141857.1 | - | 5, 6 | 411 |