Gene Transcripts

Gene Symbol
FAS
Entrez Gene ID
355
Gene Name
Fas cell surface death receptor
Gene Aliases
ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
Chromosome Location
Chr. 10: 88968429 - 89017061 on Build GRCh38
UniGene ID
Hs.244139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000043.5 NP_000034.1 2 427
NM_001320619.1 NP_001307548.1 2 427
NM_152871.3 NP_690610.1 2 427
NM_152872.3 NP_690611.1 2 427
NR_028033.3 2 427
NR_028034.3 2 427
NR_028035.3 2 427
NR_028036.3 2 427
NR_135313.1 2 427
NR_135314.1 2 418
NR_135315.1 2 418
XM_006717819.3 XP_006717882.1 3 5109
XM_011539764.2 XP_011538066.1 3 498
XM_011539765.2 XP_011538067.1 3 286
XM_011539766.2 XP_011538068.1 3 238
XM_011539767.2 XP_011538069.1 2 161
GenBank mRNA AB209361.1 - 3 3744
AK290978.1 - 2 355
AK311164.1 - 2 275
AK311424.1 - 2 418
AK311499.1 - 2 275
AV715411.1 - 2 263
AY495076.2 - 2 303
BC012479.1 - 2 419 4514272
CR450307.1 - 2 109
DA040126.1 - 2 286
DA699562.1 - 2 427
EU481974.2 - 2 303
FJ200481.1 - 2 109
FJ373045.1 - 2 109
FJ373046.1 - 2 109
FM246458.1 - 2 109
FM246459.1 - 2 109
M67454.1 - 2 303
X63717.1 - 2 329
X83490.1 - 2 134
X83491.1 - 2 134
X83492.1 - 2 134
X83493.1 - 2 134
X89101.1 - 2 109
Z47993.1 - 2 109
Z47994.1 - 2 109
Z47995.1 - 2 109
Z66556.1 - 2 109
Z70519.1 - 2 109
Z70520.1 - 2 109

View Full Product Details