Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252678.1 | NP_001239607.1 | 7 | 509 | |
NM_001320665.1 | NP_001307594.1 | 8 | 556 | ||
NM_033028.4 | NP_149017.2 | 8 | 556 | ||
NR_045565.1 | 9 | 614 | |||
NR_045566.1 | 8 | 869 | |||
XM_011521848.1 | XP_011520150.1 | 8 | 531 | ||
XM_011521849.1 | XP_011520151.1 | 7 | 419 | ||
XM_011521851.1 | XP_011520153.1 | 8 | 531 | ||
XM_017022450.1 | XP_016877939.1 | 8 | 837 | ||
XM_017022451.1 | XP_016877940.1 | 7 | 497 | ||
XM_017022452.1 | XP_016877941.1 | 7 | 727 | ||
XM_017022453.1 | XP_016877942.1 | 7 | 414 | ||
XM_017022454.1 | XP_016877943.1 | 6 | 364 | ||
GenBank mRNA | AF359281.1 | - | 8 | 528 | |
AK075321.1 | - | 8 | 520 | ||
AK298922.1 | - | 9 | 586 | ||
AK302224.1 | - | 8 | 823 | ||
AK303706.1 | - | 7 | 504 | ||
AK310106.1 | - | 2 | 67 | ||
AK314507.1 | - | 8 | 528 | ||
AY457143.1 | - | 8 | 628 | ||
BC008923.2 | - | 5 | 333 | ||
BC027624.2 | - | 8 | 526 | 4824276 | |
BE727117.1 | - | 7 | 497 | ||
BI562463.1 | - | 8 | 551 | ||
BX647855.1 | - | 7 | 480 | ||
DC372967.1 | - | 8 | 556 | ||
KU177936.1 | - | 8 | 507 |