Gene Transcripts

Gene Symbol
DIO3
Entrez Gene ID
1735
Gene Name
deiodinase, iodothyronine, type III
Gene Aliases
5DIII, D3, DIOIII, TXDI3
Chromosome Location
Chr. 14: 101561351 - 101563452 on Build GRCh38
UniGene ID
Hs.49322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001362.3 NP_001353.4 1 509
GenBank mRNA AK292310.1 - 1 365
BC017717.1 - 1 357 4395664
BX161470.1 - 1 509
S79854.1 - 1 505

View Full Product Details