Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001178096.1 | NP_001171567.1 | 2 | 411 | |
NM_001993.4 | NP_001984.1 | 2 | 411 | ||
GenBank mRNA | AF487337.1 | - | 2 | 191 | |
AF497569.1 | - | 2 | 138 | ||
AF497570.1 | - | 2 | 138 | ||
AK310918.1 | - | 3 | 805 | ||
AK312638.1 | - | 2 | 307 | ||
AY940729.1 | - | 3 | 808 | ||
AY940730.1 | - | 2 | 191 | ||
AY940731.1 | - | 2 | 191 | ||
BC011029.1 | - | 2 | 358 | 4073183 | |
BT019808.1 | - | 2 | 190 | ||
CR541792.1 | - | 2 | 190 | ||
DB203987.1 | - | 2 | 369 | ||
DB499288.1 | - | 2 | 413 | ||
J02931.1 | - | 2 | 301 | ||
M16553.1 | - | 2 | 265 | ||
M27436.1 | - | 2 | 289 |