Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289160.1 | NP_001276089.1 | 3 | 534 | |
NM_001289161.1 | NP_001276090.1 | 3 | 504 | ||
NM_005931.4 | NP_005922.2 | 3 | 633 | ||
GenBank mRNA | AF021221.1 | - | 2 | 419 | |
AF021222.1 | - | 2 | 419 | ||
AF021223.1 | - | 2 | 419 | ||
AF021224.1 | - | 2 | 419 | ||
AF021225.1 | - | 2 | 419 | ||
AF021226.1 | - | 2 | 419 | ||
AK291927.1 | - | 3 | 631 | ||
AK300791.1 | - | 3 | 534 | ||
AK303655.1 | - | 2 | 768 | ||
AK314228.1 | - | 3 | 538 | ||
AY885251.1 | - | 4 | 585 | ||
BC044218.1 | - | 3 | 468 | 6025253 | |
DC364717.1 | - | 3 | 497 | ||
GU235997.1 | - | 3 | 487 | ||
U95729.1 | - | 2 | 420 | ||
U95730.1 | - | 2 | 420 | ||
U95731.1 | - | 2 | 420 | ||
U95732.1 | - | 2 | 420 | ||
U95733.1 | - | 2 | 420 | ||
U95734.1 | - | 2 | 420 | ||
X91625.1 | - | 3 | 494 |